Assay Details
Gene Symbol: Ddb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK011756.1 | - | 2 - 3 | 1064 | 65 | |
AK158770.1 | - | 3 - 4 | 586 | 65 | ||
BC099410.1 | - | 3 - 4 | 473 | 30933658 | 65 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
