Assay Details
Gene Symbol: Erap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_030711.4 | NP_109636.1 | 12 - 13 | 1800 | 81 | |
GenBank mRNA | AB047552.1 | - | 11 - 12 | 1788 | 81 | |
AF227511.1 | - | 11 - 12 | 1776 | 81 | ||
AK029841.1 | - | 12 - 13 | 1799 | 81 | ||
AK030329.1 | - | 12 - 13 | 1801 | 81 | ||
AK036559.1 | - | 1801 | 81 | |||
AK040350.1 | - | 9 - 10 | 1018 | 81 | ||
AK159406.1 | - | 12 - 13 | 1801 | 81 | ||
BC046610.1 | - | 12 - 13 | 1806 | 6397585 | 81 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
