Assay Details
Gene Symbol: Slc12a9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031406.3 | NP_113583.2 | 10 - 11 | 1521 | 79 | |
GenBank mRNA | AF314957.1 | - | 9 - 10 | 1387 | 79 | |
AK154118.1 | - | 10 - 11 | 1480 | 79 | ||
BC046982.1 | - | 10 - 11 | 1441 | 6478046 | 79 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
