Assay Details
Gene Symbol: Arpp21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001177616.1 | NP_001171087.1 | 13 - 14 | 1664 | 63 | |
NM_001177617.1 | NP_001171088.1 | 11 - 12 | 1684 | 63 | ||
NM_001177618.1 | NP_001171089.1 | 13 - 14 | 1422 | 63 | ||
NM_033264.2 | NP_150289.1 | 14 - 15 | 1656 | 63 | ||
GenBank mRNA | AF324451.1 | - | 14 - 15 | 1616 | 63 | |
AK032417.1 | - | 13 - 14 | 1590 | 63 | ||
AK032543.1 | - | 12 - 13 | 1337 | 63 | ||
AK034376.1 | - | 12 - 13 | 1270 | 63 | ||
AK137974.1 | - | 11 - 12 | 1685 | 63 | ||
BC053001.1 | - | 12 - 13 | 1388 | 5686446 | 63 | |
BC098368.1 | - | 13 - 14 | 3888 | 63 | ||
BC128376.1 | - | 12 - 13 | 1269 | 63 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
