Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0200007465.mm.1 |
TC0200000929.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200007465.mm.1 |
NM_001242378.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200007465.mm.1 |
NM_001242378.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200007465.mm.1 |
NM_001242379.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200007465.mm.1 |
NM_001242379.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200007465.mm.1 |
NM_001242381.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200007465.mm.1 |
NM_001242380.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200007465.mm.1 |
NM_001242380.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200007465.mm.1 |
NM_001242381.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200007465.mm.1 |
NM_001242382.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200007465.mm.1 |
NM_001242382.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200007465.mm.1 |
NM_001242383.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200007465.mm.1 |
NM_001242383.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200007465.mm.1 |
NM_033552.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200007465.mm.1 |
NM_033552.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200007466.mm.1 |
TC0200000929.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200007466.mm.1 |
NM_001242378.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200007466.mm.1 |
NM_001242378.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200007466.mm.1 |
NM_001242379.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200007466.mm.1 |
NM_001242379.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200007466.mm.1 |
NM_001242380.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200007466.mm.1 |
NM_001242380.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200007466.mm.1 |
NM_001242381.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200007466.mm.1 |
NM_001242381.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200007466.mm.1 |
NM_001242382.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200007466.mm.1 |
NM_001242382.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200007466.mm.1 |
NM_001242383.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200007466.mm.1 |
NM_001242383.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200007466.mm.1 |
NM_033552.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200007466.mm.1 |
NM_033552.3 |
Refseq |