Assay Details
Gene Symbol: Slc4a10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001242378.1 | NP_001229307.1 | 23 - 24 | 3285 | 89 | |
NM_001242379.1 | NP_001229308.1 | 22 - 23 | 3195 | 89 | ||
NM_001242380.1 | NP_001229309.1 | 23 - 24 | 3285 | 89 | ||
NM_001242381.1 | NP_001229310.1 | 22 - 23 | 3192 | 89 | ||
NM_001242382.1 | NP_001229311.1 | 23 - 24 | 3282 | 89 | ||
NM_001242383.1 | NP_001229312.1 | 22 - 23 | 3192 | 89 | ||
NM_033552.3 | NP_291030.2 | 22 - 23 | 3195 | 89 | ||
GenBank mRNA | AB033759.1 | - | 22 - 23 | 3116 | 89 | |
AK043017.1 | - | 22 - 23 | 3171 | 89 | ||
AK220501.1 | - | 23 - 24 | 3250 | 89 | ||
BC039226.1 | - | 22 - 23 | 3144 | 4480841 | 89 | |
HM149278.1 | - | 22 - 23 | 3156 | 89 | ||
HQ533147.1 | - | 23 - 24 | 3222 | 89 | ||
HQ825316.1 | - | 23 - 24 | 3225 | 89 | ||
HQ831517.1 | - | 22 - 23 | 3132 | 89 | ||
JF500487.1 | - | 22 - 23 | 3272 | 89 | ||
JF500488.1 | - | 23 - 24 | 3362 | 89 | ||
JF500489.1 | - | 22 - 23 | 3272 | 89 | ||
JF500490.1 | - | 23 - 24 | 3362 | 89 | ||
JX220977.1 | - | 23 - 24 | 3249 | 89 | ||
JX254917.1 | - | 22 - 23 | 3162 | 89 | ||
JX268544.1 | - | 23 - 24 | 3363 | 89 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
