Assay Details
Gene Symbol: Smarca1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001290708.1 | NP_001277637.1 | 21 - 22 | 2948 | 82 | |
NM_053123.4 | NP_444353.3 | 21 - 22 | 2948 | 82 | ||
GenBank mRNA | AF325920.1 | - | 21 - 22 | 2958 | 82 | |
AK030741.1 | - | 21 - 22 | 2915 | 82 | ||
BC016071.1 | - | 21 - 22 | 2925 | 82 | ||
BC057115.1 | - | 21 - 22 | 2948 | 6406330 | 82 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
