Assay Details
Gene Symbol: Rad51c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001291440.1 | NP_001278369.1 | 3 - 4 | 781 | 154 | |
NM_053269.3 | NP_444499.1 | 3 - 4 | 613 | 154 | ||
GenBank mRNA | AK076551.1 | - | 3 - 4 | 781 | 154 | |
BC090648.1 | - | 557 | 30640414 | 154 | ||
BC141034.1 | - | 3 - 4 | 608 | 9055646 | 154 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
