Assay Details
Gene Symbol: Rad51c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001291440.1 | NP_001278369.1 | 5 - 6 | 1049 | 107 | |
NM_053269.3 | NP_444499.1 | 5 - 6 | 881 | 107 | ||
GenBank mRNA | AK076551.1 | - | 5 - 6 | 1049 | 107 | |
AK136204.1 | - | 5 - 6 | 895 | 107 | ||
BC090648.1 | - | 825 | 30640414 | 107 | ||
BC141034.1 | - | 5 - 6 | 876 | 9055646 | 107 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
