Assay Details
Gene Symbol: Mta1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_054081.2 | NP_473422.2 | 18 - 19 | 1935 | 66 | |
GenBank mRNA | AB794093.1 | - | 19 - 20 | 1846 | 66 | |
AB794094.1 | - | 18 - 19 | 1795 | 66 | ||
AB794095.1 | - | 19 - 20 | 1799 | 66 | ||
AB794097.1 | - | 18 - 19 | 1748 | 66 | ||
AB794100.1 | - | 19 - 20 | 1760 | 66 | ||
AB794101.1 | - | 19 - 20 | 1891 | 66 | ||
AB794102.1 | - | 18 - 19 | 1735 | 66 | ||
AB794103.1 | - | 19 - 20 | 1807 | 66 | ||
AF288137.1 | - | 18 - 19 | 1897 | 66 | ||
AF463504.1 | - | 19 - 20 | 1846 | 66 | ||
BC005698.1 | - | 7 - 8 | 704 | 66 | ||
BC024527.1 | - | 11 - 12 | 1161 | 66 | ||
BC025167.1 | - | 7 - 8 | 692 | 66 | ||
CX213009.1 | - | 5 - 6 | 380 | 66 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
