Assay Details
Gene Symbol: Anpep | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008486.2 | NP_032512.2 | 5 - 6 | 1272 | 67 | |
GenBank mRNA | AK143746.1 | - | 5 - 6 | 1215 | 67 | |
AK171427.1 | - | 6 - 7 | 1634 | 67 | ||
BC005431.1 | - | 5 - 6 | 1238 | 3593307 | 67 | |
BC017011.1 | - | 5 - 6 | 1296 | 3590117 | 67 | |
BC040792.1 | - | 5 - 6 | 1296 | 3590117 | 67 | |
U77083.1 | - | 5 - 6 | 1204 | 67 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
