Assay Details
Gene Symbol: Pde6b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008806.2 | NP_032832.2 | 15 - 16 | 1988 | 70 | |
GenBank mRNA | AK039397.1 | - | 15 - 16 | 1700 | 70 | |
AK044364.1 | - | 15 - 16 | 1931 | 70 | ||
BC048247.1 | - | 15 - 16 | 1967 | 70 | ||
BC058803.1 | - | 13 - 14 | 2847 | 70 | ||
BC129923.1 | - | 15 - 16 | 1918 | 40111882 | 70 | |
BC129924.1 | - | 15 - 16 | 1917 | 70 | ||
X55968.1 | - | 15 - 16 | 1921 | 70 | ||
X60133.1 | - | 15 - 16 | 1928 | 70 | ||
X87952.1 | - | 15 - 16 | 1928 | 70 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
