Assay Details
Gene Symbol: Prim1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008921.2 | NP_032947.1 | 12 - 13 | 1266 | 83 | |
GenBank mRNA | AK011456.1 | - | 12 - 13 | 1248 | 83 | |
AK088557.1 | - | 12 - 13 | 1286 | 83 | ||
AK133878.1 | - | 12 - 13 | 1263 | 83 | ||
AK160831.1 | - | 10 - 11 | 3028 | 83 | ||
AK169632.1 | - | 12 - 13 | 1264 | 83 | ||
BC089544.1 | - | 12 - 13 | 1305 | 30504228 | 83 | |
D13544.1 | - | 12 - 13 | 1266 | 83 | ||
J04620.1 | - | 12 - 13 | 1325 | 83 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
DNA replication
DNA replication, synthesis of RNA primer
transcription, DNA-templated
telomerase activity
RNA helicase activity
DNA primase activity
DNA-directed RNA polymerase activity
sulfate adenylyltransferase activity
protein binding
RNA guanylyltransferase activity
tRNA guanylyltransferase activity
mannose-phosphate guanylyltransferase activity
transferase activity
nucleotidyltransferase activity
DNA/RNA helicase activity
CTP:2,3-di-O-geranylgeranyl-sn-glycero-1-phosphate cytidyltransferase activity
phospholactate guanylyltransferase activity
ATP:coenzyme F420 adenylyltransferase activity
metal ion binding
UDP-N-acetylgalactosamine diphosphorylase activity
DNA replication
DNA replication, synthesis of RNA primer
transcription, DNA-templated
telomerase activity
RNA helicase activity
DNA primase activity
DNA-directed RNA polymerase activity
sulfate adenylyltransferase activity
protein binding
RNA guanylyltransferase activity
tRNA guanylyltransferase activity
mannose-phosphate guanylyltransferase activity
transferase activity
nucleotidyltransferase activity
DNA/RNA helicase activity
CTP:2,3-di-O-geranylgeranyl-sn-glycero-1-phosphate cytidyltransferase activity
phospholactate guanylyltransferase activity
ATP:coenzyme F420 adenylyltransferase activity
metal ion binding
UDP-N-acetylgalactosamine diphosphorylase activity
MicroArray Probes:
Agilent
Affymetrix
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P168632 | D13544.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P168632 | J04620.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P168632 | AK133878.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P168632 | AK011456.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P168632 | AK088557.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P168632 | D13544.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P168632 | J04620.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P168632 | AK133878.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P168632 | AK011456.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P168632 | AK088557.1 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 478749 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 342883 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 453735 | AK090057.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 487665 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 816061 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 452701 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 84122 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 179415 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 730286 | AB290927.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 730286 | AF271381.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 730286 | AK046772.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 730286 | BC046386.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 730286 | BC116297.2 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 730286 | BC116323.2 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 730286 | NM_181422.2 | refseq_mrna_mouse_NM |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 836641 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 103584 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 84094 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P159754 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P112322 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P168632 | D13544.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P168632 | J04620.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P168632 | AK133878.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P168632 | AK011456.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P168632 | AK088557.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P168632 | D13544.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P168632 | J04620.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P168632 | AK133878.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P168632 | AK011456.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P168632 | AK088557.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P168632 | D13544.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P168632 | J04620.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P168632 | AK133878.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P168632 | AK011456.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P168632 | AK088557.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P168632 | D13544.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P168632 | J04620.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P168632 | AK133878.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P168632 | AK011456.1 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P168632 | AK088557.1 | mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27539079 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27539084 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27539080 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27539081 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27539074 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27539076 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27539082 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27539078 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P27539071 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P07231889 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P159754 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P07231856 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P112322 | - | - |
Whole Mouse Genome G4122A | A_51_P168632 | D13544.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P168632 | J04620.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P168632 | AK133878.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P168632 | AK011456.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P168632 | AK088557.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P168632 | D13544.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P168632 | J04620.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P168632 | AK133878.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P168632 | AK011456.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P168632 | AK088557.1 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P131581 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P159754 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P112322 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08964297 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08964301 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27539071 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27539108 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27539156 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27539031 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27539090 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27539111 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27539172 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27539178 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27539181 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27539076 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27539040 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27539043 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27539101 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27539127 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27539180 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27539093 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27539189 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27539116 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27539133 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27539168 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27539119 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27539153 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27539175 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27539131 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27539080 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27539081 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27539082 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27539109 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27539094 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27539135 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27539110 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27539161 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27539100 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27539037 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27539184 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27539150 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27539166 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27539102 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27539164 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27539122 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27539179 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27539130 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27539106 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27539104 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27539050 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27539162 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27539159 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27539033 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27539145 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27539182 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27539124 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27539171 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27539107 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27539088 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27539098 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27539152 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27539137 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27539120 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27539139 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27539036 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27539186 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27539143 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27539035 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27539163 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27539070 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P27539155 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27539069 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27539085 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27539065 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27539094 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27539079 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27539075 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P08634267 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P08280001 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P08964297 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27539084 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P08910543 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27539096 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27539128 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27539102 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27539090 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27539078 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27539081 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27539097 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27539068 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P08463055 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27539083 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27539095 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27539080 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27539066 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27539103 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27539093 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27539133 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P08227325 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P08309586 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27539071 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27539088 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27539076 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27539082 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27539098 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P08964302 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P27539131 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10367089 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10367077 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10367079 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10367085 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10367086 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10367087 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10367081 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10367090 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10367080 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10367088 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10367083 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10367082 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10367084 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10367078 | - | - |
Affymetrix HT MG-430 PM Array Plate | 1418369_PM_at | - | - |
Affymetrix HT MG-430 PM Array Plate | 1449061_PM_a_at | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10367076 | - | - |
Mouse Genome 430 2.0 Array | 1449061_a_at | - | - |
Mouse Genome 430 2.0 Array | 1444544_at | - | - |
Mouse Genome 430 2.0 Array | 1444544_PM_at | - | - |
Mouse Genome 430 2.0 Array | 1449061_PM_a_at | - | - |
Mouse Genome 430 2.0 Array | 1418369_at | - | - |
Mouse Genome 430 2.0 Array | 1418369_PM_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 19075_PM_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 19075_at | - | - |
Affymetrix Murine Genome 6500A Array | Msa1530_at | - | - |
Affymetrix Mouse Clariom S Array | PSR1000011419.mm.1 | NM_008921.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1000011419.mm.1 | NM_008921.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1000011419.mm.1 | TC1000001579.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1000011416.mm.1 | NM_008921.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1000011416.mm.1 | NM_008921.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1000011416.mm.1 | TC1000001579.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1000011418.mm.1 | NM_008921.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1000011418.mm.1 | NM_008921.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1000011418.mm.1 | TC1000001579.mm.1 | MTA TC |
Mouse Expression Set 430 | 1418369_at | - | - |
Mouse Expression Set 430 | 1449061_a_at | - | - |
Mouse Expression Set 430 | 1449061_PM_a_at | - | - |
Mouse Expression Set 430 | 1418369_PM_at | - | - |
Murine Genome U74v2 Set | 96772_at | - | - |
Murine Genome U74v2 Set | 96772_PM_at | - | - |
Mouse Genome 430A 2.0 Array | 1418369_PM_at | - | - |
Mouse Genome 430A 2.0 Array | 1449061_a_at | - | - |
Mouse Genome 430A 2.0 Array | 1418369_at | - | - |
Mouse Genome 430A 2.0 Array | 1449061_PM_a_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1449061_PM_a_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1418369_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1418369_PM_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1449061_a_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina mouse-6 v1.0 expression beadchip | 6420746 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0006420746 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2743208 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_2743208 | - | - |
mouse-6 v1.1 (Illumina) | scl38302.14.1_13-S | - | - |
Illumina Sentrix mouse-6.v1 bead array | 6420746 | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl38302.14.1_13-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 6420746 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_1241140 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1241140 | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl38302.14.1_13-S | - | - |
