Assay Details
Gene Symbol: Adam19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001291890.1 | NP_001278819.1 | 4 - 5 | 275 | 69 | |
NM_001291891.1 | NP_001278820.1 | 4 - 5 | 460 | 69 | ||
NM_009616.4 | NP_033746.1 | 4 - 5 | 460 | 69 | ||
GenBank mRNA | AF019887.2 | - | 4 - 5 | 365 | 69 | |
AK147217.1 | - | 4 - 5 | 383 | 69 | ||
AK147549.1 | - | 4 - 5 | 276 | 69 | ||
BC093480.1 | - | 4 - 5 | 451 | 69 | ||
CJ114671.1 | - | 4 - 5 | 69 | |||
D50410.2 | - | 4 - 5 | 392 | 69 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001291891.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
