Assay Details
Gene Symbol: Ahrr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009644.2 | NP_033774.1 | 9 - 10 | 1038 | 78 | |
GenBank mRNA | AB015140.1 | - | 9 - 10 | 1038 | 78 | |
AK154152.1 | - | 9 - 10 | 1027 | 78 | ||
AK154761.1 | - | 9 - 10 | 1060 | 78 | ||
AK155711.1 | - | 9 - 10 | 1060 | 78 | ||
BC118534.1 | - | 8 - 9 | 844 | 40095111 | 78 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
