Assay Details
Gene Symbol: Akr1b7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009731.2 | NP_033861.2 | 4 - 5 | 484 | 96 | |
GenBank mRNA | AK002705.1 | - | 4 - 5 | 486 | 96 | |
AK147775.1 | - | 4 - 5 | 513 | 96 | ||
BC086929.1 | - | 4 - 5 | 457 | 6775941 | 96 | |
J05663.1 | - | 4 - 5 | 456 | 96 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
