Assay Details
Gene Symbol: Bmpr1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009758.4 | NP_033888.2 | 11 - 12 | 1746 | 80 | |
GenBank mRNA | AK132051.1 | - | 11 - 12 | 1759 | 80 | |
AK143804.1 | - | 2 - 3 | 373 | 80 | ||
AY365062.1 | - | 9 - 10 | 1340 | 80 | ||
BC042611.1 | - | 10 - 11 | 1536 | 5364272 | 80 | |
D16250.1 | - | 9 - 10 | 1481 | 80 | ||
U04672.1 | - | 9 - 10 | 1350 | 80 | ||
Z23154.1 | - | 9 - 10 | 1340 | 80 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
