Assay Details
Gene Symbol: Per2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011066.3 | NP_035196.2 | 14 - 15 | 1797 | 106 | |
GenBank mRNA | AF035830.1 | - | 14 - 15 | 1744 | 106 | |
AF036893.1 | - | 14 - 15 | 1767 | 106 | ||
AK122253.1 | - | 14 - 15 | 1762 | 106 | ||
AK159847.1 | - | 14 - 15 | 1799 | 106 | ||
AK165556.1 | - | 14 - 15 | 1800 | 106 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
