Assay Details
Gene Symbol: Anxa11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013469.2 | NP_038497.2 | 13 - 14 | 1477 | 66 | |
GenBank mRNA | AK137158.1 | - | 12 - 13 | 1787 | 66 | |
AK140375.1 | - | 13 - 14 | 1450 | 66 | ||
BC012875.1 | - | 13 - 14 | 1397 | 3500318 | 66 | |
CT010331.1 | - | 12 - 13 | 1328 | 66 | ||
U65986.1 | - | 13 - 14 | 1462 | 66 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
