Assay Details
Gene Symbol: Phtf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001163467.1 | NP_001156939.1 | 13 - 14 | 1817 | 79 | |
NM_001163468.1 | NP_001156940.1 | 12 - 13 | 2064 | 79 | ||
NM_001163469.1 | NP_001156941.1 | 12 - 13 | 2088 | 79 | ||
NM_013629.2 | NP_038657.2 | 13 - 14 | 2223 | 79 | ||
GenBank mRNA | AJ133721.1 | - | 13 - 14 | 1817 | 79 | |
AJ242864.1 | - | 13 - 14 | 2223 | 79 | ||
AK019146.1 | - | 13 - 14 | 2127 | 79 | ||
AK088714.1 | - | 12 - 13 | 1955 | 79 | ||
AK131629.1 | - | 2114 | 79 | |||
BC021147.1 | - | 13 - 14 | 2096 | 79 | ||
BC050825.1 | - | 12 - 13 | 1900 | 6518647 | 79 | |
BU053202.1 | - | 1 - 2 | 223 | 79 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
