Assay Details
Gene Symbol: Psmb10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013640.3 | NP_038668.2 | 3 - 4 | 627 | 74 | |
GenBank mRNA | AK144096.1 | - | 3 - 4 | 350 | 74 | |
BC004730.1 | - | 3 - 4 | 353 | 3583052 | 74 | |
CT010274.1 | - | 3 - 4 | 245 | 74 | ||
D85561.1 | - | 3 - 4 | 322 | 74 | ||
U77784.1 | - | 3 - 4 | 314 | 74 | ||
Y10875.1 | - | 3 - 4 | 345 | 74 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
cell morphogenesis
proteolysis
proteasomal ubiquitin-independent protein catabolic process
T cell proliferation
proteasome-mediated ubiquitin-dependent protein catabolic process
proteolysis involved in cellular protein catabolic process
endopeptidase activity
threonine-type endopeptidase activity
peptidase activity
hydrolase activity
cell morphogenesis
proteolysis
proteasomal ubiquitin-independent protein catabolic process
T cell proliferation
proteasome-mediated ubiquitin-dependent protein catabolic process
proteolysis involved in cellular protein catabolic process
endopeptidase activity
threonine-type endopeptidase activity
peptidase activity
hydrolase activity
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P570266 | U77784.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P570266 | Y10875.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P570266 | D85561.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P570266 | NM_013640.3 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P570266 | AK144096.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P570266 | U77784.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P570266 | Y10875.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P570266 | D85561.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P570266 | NM_013640.3 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P570266 | AK144096.1 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 780872 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 969672 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P143911 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P570266 | U77784.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P570266 | Y10875.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P570266 | D85561.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P570266 | NM_013640.3 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P570266 | AK144096.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P570266 | U77784.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P570266 | Y10875.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P570266 | D85561.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P570266 | NM_013640.3 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P570266 | AK144096.1 | mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26136859 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26136871 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26136853 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26136848 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26136856 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26136851 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26136873 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26136865 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26136847 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26136854 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26136863 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26136868 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26136855 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26136860 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26136850 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26136857 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26136869 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26136852 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P143911 | - | - |
Whole Mouse Genome G4122A | A_52_P570266 | U77784.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P570266 | Y10875.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P570266 | D85561.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P570266 | NM_013640.3 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P570266 | AK144096.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P570266 | U77784.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P570266 | Y10875.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P570266 | D85561.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P570266 | NM_013640.3 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P570266 | AK144096.1 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P113098 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P143911 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06977306 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06977326 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06977339 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P06977337 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26136836 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26136848 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26136855 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26136826 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26136831 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26136856 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26136898 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26136889 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26136886 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26136834 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26136824 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26136833 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26136843 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26136821 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26136847 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26136832 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26136857 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26136868 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26136873 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26136885 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26136854 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26136815 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26136892 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26136859 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26136900 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26136863 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26136890 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26136853 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26136865 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26136817 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26136897 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26136860 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26136818 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26136884 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26136888 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26136841 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26136829 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26136896 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26136895 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26136822 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26136839 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26136851 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26136857 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26136863 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26136855 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26136873 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26136860 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26136869 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26136853 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26136850 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26136868 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26136872 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26136848 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26136847 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26136870 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06911458 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26136856 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26136852 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26136859 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26136854 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P06977336 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26136871 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10581382 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10581384 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10581386 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10581383 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10581379 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10581381 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10581380 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10581387 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10581385 | - | - |
Affymetrix HT MG-430 PM Array Plate | 1448632_PM_at | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10581378 | - | - |
Mouse Genome 430 2.0 Array | 1448632_PM_at | - | - |
Mouse Genome 430 2.0 Array | 1448632_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 19171_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 19171_PM_at | - | - |
Affymetrix Mouse Clariom S Array | PSR0800021886.mm.1 | NM_013640.3.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0800021886.mm.1 | NM_013640.3 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0800021886.mm.1 | TC0800002842.mm.1 | MTA TC |
Mouse Expression Set 430 | 1448632_PM_at | - | - |
Mouse Expression Set 430 | 1448632_at | - | - |
Murine Genome U74v2 Set | 101486_PM_at | - | - |
Murine Genome U74v2 Set | 101486_at | - | - |
Mouse Genome 430A 2.0 Array | 1448632_at | - | - |
Mouse Genome 430A 2.0 Array | 1448632_PM_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1448632_PM_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1448632_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 925595 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
MouseWG-6 v2 Expression BeadChip | 630504 | - | - |
MouseWG-6 v2 Expression BeadChip | 6510039 | - | - |
Illumina mouse-6 v1.0 expression beadchip | 6510039 | - | - |
Illumina mouse-6 v1.0 expression beadchip | 630504 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0006510039 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0000630504 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1259510 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2757870 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_2757870 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_1259510 | - | - |
mouse-6 v1.1 (Illumina) | scl34384.4.1_27-S | - | - |
mouse-6 v1.1 (Illumina) | scl0019171.2_204-S | - | - |
Illumina Sentrix mouse-6.v1 bead array | 6510039 | - | - |
Illumina Sentrix mouse-6.v1 bead array | 630504 | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl0019171.2_204-S | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl34384.4.1_27-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 6510039 | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 630504 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_2914843 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_2757870 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2757870 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2914843 | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl0019171.2_204-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl34384.4.1_27-S | - | - |
