Assay Details
Gene Symbol: Srcin1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_018873.2 | NP_061361.2 | 17 - 18 | 3361 | 71 | |
GenBank mRNA | AF040944.1 | - | 17 - 18 | 3363 | 71 | |
AK129422.1 | - | 12 - 13 | 2518 | 71 | ||
U59873.1 | - | 17 - 18 | 3362 | 71 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
