Assay Details
Gene Symbol: Net1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019671.3 | NP_062645.2 | 1 - 2 | 283 | 139 | |
GenBank mRNA | AF094520.1 | - | 1 - 2 | 179 | 139 | |
CF167482.1 | - | 1 - 2 | 267 | 139 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
