Assay Details
Gene Symbol: Ncoa6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019825.3 | NP_062799.2 | 10 - 11 | 6049 | 93 | |
GenBank mRNA | AF216186.1 | - | 9 - 10 | 5934 | 93 | |
AK129079.1 | - | 3 - 4 | 3208 | 93 | ||
AK142566.1 | - | 4 - 5 | 4234 | 93 | ||
BC083187.1 | - | 10 - 11 | 6049 | 30544739 | 93 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
