Assay Details
Gene Symbol: Brd4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001286630.1 | NP_001273559.1 | 8 - 9 | 1652 | 102 | |
NM_020508.4 | NP_065254.3 | 8 - 9 | 1605 | 102 | ||
NM_198094.2 | NP_932762.2 | 8 - 9 | 1595 | 102 | ||
GenBank mRNA | AF273217.1 | - | 7 - 8 | 1520 | 102 | |
AF461395.1 | - | 8 - 9 | 1581 | 102 | ||
AF461396.1 | - | 8 - 9 | 1581 | 102 | ||
AK042431.1 | - | 9 - 10 | 1823 | 102 | ||
AK147545.1 | - | 8 - 9 | 1617 | 102 | ||
AK154514.1 | - | 7 - 8 | 1580 | 102 | ||
BC011541.1 | - | 7 - 8 | 1441 | 102 | ||
BC082782.1 | - | 7 - 8 | 1393 | 102 | ||
BC103773.1 | - | 7 - 8 | 1519 | 102 | ||
BC138834.1 | - | 8 - 9 | 1575 | 8861856 | 102 | |
BC138835.1 | - | 8 - 9 | 1574 | 8861857 | 102 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
