Assay Details
Gene Symbol: Nrgn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022029.2 | NP_071312.1 | 1 - 2 | 208 | 63 | |
GenBank mRNA | AK002933.1 | - | 1 - 2 | 168 | 63 | |
AK158630.1 | - | 1 - 2 | 195 | 63 | ||
BC061102.1 | - | 1 - 2 | 172 | 6590375 | 63 | |
BC138511.1 | - | 1 - 2 | 85 | 8861531 | 63 | |
BC138513.1 | - | 1 - 2 | 85 | 8861533 | 63 | |
BU604757.1 | - | 1 - 2 | 211 | 63 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the AK002933.1, AK158630.1, BC061102.1, BC138511.1, BC138513.1, NM_022029.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
