Assay Details
Gene Symbol: Cenpm | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001080158.1 | NP_001073627.1 | 4 - 5 | 468 | 96 | |
NM_025639.4 | NP_079915.1 | 5 - 6 | 469 | 96 | ||
GenBank mRNA | AK010885.1 | - | 5 - 6 | 461 | 96 | |
AK011191.1 | - | 5 - 6 | 446 | 96 | ||
AK011227.1 | - | 5 - 6 | 453 | 96 | ||
AK011464.1 | - | 5 - 6 | 470 | 96 | ||
AK011755.1 | - | 5 - 6 | 462 | 96 | ||
AK011793.1 | - | 5 - 6 | 455 | 96 | ||
AK017679.1 | - | 5 - 6 | 462 | 96 | ||
AY243536.1 | - | 5 - 6 | 430 | 96 | ||
AY243537.1 | - | 5 - 6 | 430 | 96 | ||
BG276334.1 | - | 4 - 5 | 468 | 96 | ||
BY710734.1 | - | 5 - 6 | 470 | 96 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30382331 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30382334 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30382333 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30382336 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30382329 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30382332 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30382330 | - | - |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P463087 | BC009160.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P463087 | BG276334.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P463087 | BY710734.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P463087 | AK011755.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P463087 | AK011793.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P463087 | BC009160.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P463087 | BG276334.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P463087 | BY710734.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P463087 | AK011755.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P463087 | AK011793.1 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 817684 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 660494 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 485616 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 2291 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 49126 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 831452 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 206260 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 897604 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 151009 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P463087 | BC009160.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P463087 | BG276334.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P463087 | BY710734.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P463087 | AK011755.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P463087 | AK011793.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P463087 | BC009160.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P463087 | BG276334.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P463087 | BY710734.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P463087 | AK011755.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P463087 | AK011793.1 | mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30382340 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30382332 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30382348 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30382342 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30382344 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30382335 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30382346 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30382337 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30382325 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30382326 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30382336 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30382323 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30382329 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30382341 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30382338 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30382330 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P08437548 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P08437540 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P03083786 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P08437567 | - | - |
Whole Mouse Genome G4122A | A_51_P463087 | BC009160.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P463087 | BG276334.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P463087 | BY710734.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P463087 | AK011755.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P463087 | AK011793.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P463087 | BC009160.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P463087 | BG276334.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P463087 | BY710734.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P463087 | AK011755.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P463087 | AK011793.1 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P102105 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P03083786 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P12346237 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30382334 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30382330 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30382332 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30382333 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30382329 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30382331 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P30382336 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30382215 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30382315 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30382242 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30382289 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30382271 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30382332 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30382218 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30382204 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30382257 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30382362 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30382207 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30382296 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30382220 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30382321 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30382228 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30382305 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30382353 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30382268 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30382256 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30382283 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30382237 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30382318 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30382282 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30382360 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30382310 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30382273 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30382259 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30382263 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30382253 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30382278 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30382277 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30382335 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30382249 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30382246 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30382244 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30382285 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30382262 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30382344 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30382281 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30382254 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30382208 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30382279 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30382213 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30382255 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30382341 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30382240 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30382337 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30382302 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30382211 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30382223 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30382205 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30382314 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30382340 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30382329 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30382358 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30382264 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30382245 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30382248 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30382227 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30382346 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30382226 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30382221 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30382225 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30382235 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30382209 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30382292 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30382336 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30382366 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30382275 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30382299 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30382352 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30382206 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30382357 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30382239 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30382345 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30382360 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30382357 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30382362 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30382346 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30382312 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30382364 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30382289 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30382310 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P11875401 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30382335 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30382353 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30382358 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30382294 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30382292 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30382338 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30382326 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30382305 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30382336 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30382340 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P11891272 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30382299 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30382344 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30382363 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30382366 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30382302 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30382322 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30382318 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P12355118 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30382348 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30382308 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30382341 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30382296 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30382317 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30382337 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30382325 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P11964951 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30382329 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30382327 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30382315 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P30382314 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P11959317 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P12124821 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10430828 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10430831 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10430833 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10430826 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10430827 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10430832 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10430830 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10430829 | - | - |
Affymetrix HT MG-430 PM Array Plate | 1418184_PM_at | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10430825 | - | - |
Mouse Genome 430 2.0 Array | 1418184_PM_at | - | - |
Mouse Genome 430 2.0 Array | 1418184_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 66570_PM_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 66570_at | - | - |
Affymetrix Mouse Clariom S Array | PSR1500015279.mm.1 | NM_001080158.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1500015279.mm.1 | NM_001080158.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1500015279.mm.1 | NM_025639.4.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1500015279.mm.1 | NM_025639.4 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1500015279.mm.1 | NM_178269.3.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1500015279.mm.1 | NM_178269.3 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1500015279.mm.1 | TC1500001940.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1500015280.mm.1 | NM_001080158.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1500015280.mm.1 | NM_001080158.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1500015280.mm.1 | NM_025639.4.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1500015280.mm.1 | NM_025639.4 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1500015280.mm.1 | NM_178269.3.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1500015280.mm.1 | NM_178269.3 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1500015280.mm.1 | TC1500001940.mm.1 | MTA TC |
Mouse Expression Set 430 | 1418184_at | - | - |
Mouse Expression Set 430 | 1418184_PM_at | - | - |
Mouse Genome 430A 2.0 Array | 1418184_PM_at | - | - |
Mouse Genome 430A 2.0 Array | 1418184_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1418184_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1418184_PM_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
MouseWG-6 v2 Expression BeadChip | 630280 | - | - |
Illumina mouse-6 v1.0 expression beadchip | 630280 | - | - |
Illumina mouse-6 v1.0 expression beadchip | 3840068 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0003840068 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0000630280 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2734882 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2733472 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_2734882 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_2733472 | - | - |
mouse-6 v1.1 (Illumina) | scl46921.7.4_52-S | - | - |
mouse-6 v1.1 (Illumina) | scl0002535.1_10-S | - | - |
Illumina Sentrix mouse-6.v1 bead array | 3840068 | - | - |
Illumina Sentrix mouse-6.v1 bead array | 630280 | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl46921.7.4_52-S | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl0002535.1_10-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 630280 | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 3840068 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_2734882 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_3104928 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_2733472 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2733472 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_3104928 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2734882 | - | - |
