Assay Details
Gene Symbol: Lonp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_025827.3 | NP_080103.1 | 4 - 5 | 863 | 105 | |
GenBank mRNA | AK004843.1 | - | 4 - 5 | 777 | 105 | |
AK050231.1 | - | 4 - 5 | 772 | 105 | ||
AK088348.1 | - | 4 - 5 | 864 | 105 | ||
AK151103.1 | - | 4 - 5 | 809 | 105 | ||
AK168791.1 | - | 4 - 5 | 809 | 105 | ||
AK168956.1 | - | 4 - 5 | 809 | 105 | ||
AK169656.1 | - | 777 | 105 | |||
AK218948.1 | - | 1 - 2 | 128 | 105 | ||
BC049090.1 | - | 4 - 5 | 799 | 6401542 | 105 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
