Assay Details
Gene Symbol: Lonp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001168591.1 | NP_001162063.1 | 2 - 3 | 217 | 133 | |
NM_025827.3 | NP_080103.1 | 8 - 9 | 1521 | 133 | ||
GenBank mRNA | AK004843.1 | - | 8 - 9 | 1435 | 133 | |
AK050231.1 | - | 8 - 9 | 1430 | 133 | ||
AK075719.1 | - | 2 - 3 | 218 | 133 | ||
AK088348.1 | - | 7 - 8 | 1427 | 133 | ||
AK151103.1 | - | 8 - 9 | 1467 | 133 | ||
AK151406.1 | - | 6 - 7 | 1035 | 133 | ||
AK152964.1 | - | 6 - 7 | 134 | |||
AK168628.1 | - | 6 - 7 | 1045 | 133 | ||
AK168791.1 | - | 8 - 9 | 1467 | 133 | ||
AK168956.1 | - | 8 - 9 | 1467 | 133 | ||
AK169656.1 | - | 1435 | 133 | |||
BC049090.1 | - | 8 - 9 | 1457 | 6401542 | 133 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
