Assay Details
Gene Symbol: Lonp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001168591.1 | NP_001162063.1 | 5 - 6 | 635 | 89 | |
NM_025827.3 | NP_080103.1 | 11 - 12 | 1939 | 89 | ||
GenBank mRNA | AK004843.1 | - | 11 - 12 | 1853 | 89 | |
AK050231.1 | - | 11 - 12 | 1848 | 89 | ||
AK075719.1 | - | 5 - 6 | 636 | 89 | ||
AK088348.1 | - | 10 - 11 | 1845 | 89 | ||
AK151103.1 | - | 11 - 12 | 1885 | 89 | ||
AK168628.1 | - | 9 - 10 | 1463 | 89 | ||
AK168791.1 | - | 11 - 12 | 1885 | 89 | ||
AK168956.1 | - | 11 - 12 | 1885 | 89 | ||
AK169656.1 | - | 1853 | 89 | |||
BC049090.1 | - | 11 - 12 | 1875 | 6401542 | 89 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
