Assay Details
Gene Symbol: Cbx4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_007625.2 | NP_031651.2 | 2 - 3 | 332 | 79 | |
GenBank mRNA | BC018304.1 | - | 2 - 3 | 332 | 79 | |
BC045162.1 | - | 2 - 3 | 316 | 79 | ||
BC117801.1 | - | 2 - 3 | 206 | 40092282 | 79 | |
BC117802.1 | - | 2 - 3 | 206 | 40092283 | 79 | |
U63387.1 | - | 2 - 3 | 244 | 79 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the BC117802.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
