Assay Details
Gene Symbol: Cdh16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001252627.1 | NP_001239556.1 | 12 - 13 | 1968 | 62 | |
NM_001252628.1 | NP_001239557.1 | 13 - 14 | 2058 | 62 | ||
NM_007663.3 | NP_031689.1 | 13 - 14 | 2058 | 62 | ||
GenBank mRNA | AF016271.1 | - | 13 - 14 | 1926 | 62 | |
AJ609635.1 | - | 13 - 14 | 1925 | 62 | ||
AK052636.1 | - | 13 - 14 | 1861 | 62 | ||
AK093905.1 | - | 13 - 14 | 1856 | 62 | ||
AK164546.1 | - | 12 - 13 | 1969 | 62 | ||
BC015251.1 | - | 13 - 14 | 1906 | 4235757 | 62 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
