Assay Details
Gene Symbol: Cdh2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_007664.4 | NP_031690.3 | 13 - 14 | 2860 | 75 | |
GenBank mRNA | AB008811.1 | - | 13 - 14 | 2537 | 75 | |
AK032826.1 | - | 13 - 14 | 2300 | 75 | ||
AK146981.1 | - | 13 - 14 | 3057 | 75 | ||
BC022107.1 | - | 13 - 14 | 2291 | 5100313 | 75 | |
M31131.1 | - | 13 - 14 | 2537 | 75 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
