Assay Details
Gene Symbol: Inadl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001005787.1 | NP_001005787.1 | 10 - 11 | 1441 | 72 | |
NM_007704.2 | NP_031730.1 | 10 - 11 | 1441 | 72 | ||
NM_172696.2 | NP_766284.2 | 36 - 37 | 4891 | 72 | ||
GenBank mRNA | AF060539.1 | - | 10 - 11 | 1341 | 72 | |
AK077268.1 | - | 23 - 24 | 3173 | 72 | ||
BC037607.1 | - | 10 - 11 | 1113 | 72 | ||
BC057124.1 | - | 10 - 11 | 1120 | 72 | ||
BC082787.1 | - | 25 - 26 | 3284 | 72 | ||
BC141404.1 | - | 36 - 37 | 4867 | 9056017 | 72 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
