Assay Details
Gene Symbol: Acsl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK004897.1 | - | 12 - 13 | 1212 | 74 | |
AK145900.1 | - | 12 - 13 | 1339 | 74 | ||
AK149406.1 | - | 12 - 13 | 1217 | 74 | ||
AK152772.1 | - | 12 - 13 | 1217 | 74 | ||
AK153050.1 | - | 12 - 13 | 1345 | 74 | ||
AK161189.1 | - | 12 - 13 | 1191 | 74 | ||
AK168078.1 | - | 12 - 13 | 1217 | 74 | ||
BC056644.1 | - | 12 - 13 | 1201 | 5098645 | 74 | |
U15977.1 | - | 11 - 12 | 1130 | 74 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
MicroArray Probes:
