Assay Details
Gene Symbol: Acsl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK004897.1 | - | 16 - 17 | 1606 | 85 | |
AK145900.1 | - | 16 - 17 | 1733 | 85 | ||
AK149406.1 | - | 16 - 17 | 1611 | 85 | ||
AK152772.1 | - | 16 - 17 | 1611 | 85 | ||
AK153050.1 | - | 16 - 17 | 1739 | 85 | ||
AK161189.1 | - | 16 - 17 | 1585 | 85 | ||
AK168078.1 | - | 16 - 17 | 1611 | 85 | ||
AK180785.1 | - | 1 - 2 | 106 | 85 | ||
BC056644.1 | - | 16 - 17 | 1595 | 5098645 | 85 | |
U15977.1 | - | 15 - 16 | 1524 | 85 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
