Assay Details
Gene Symbol: Fxr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001113188.1 | NP_001106659.1 | 4 - 5 | 299 | 70 | |
NM_001113189.1 | NP_001106660.1 | 4 - 5 | 299 | 70 | ||
NM_008053.2 | NP_032079.1 | 4 - 5 | 299 | 70 | ||
GenBank mRNA | BC019139.1 | - | 4 - 5 | 280 | 5041635 | 70 |
CJ086949.1 | - | 4 - 5 | 300 | 70 | ||
X90875.1 | - | 4 - 5 | 278 | 70 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
