Assay Details
Gene Symbol: Ganab | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001293621.1 | NP_001280550.1 | 2 - 3 | 214 | 88 | |
NM_008060.2 | NP_032086.1 | 2 - 3 | 214 | 88 | ||
GenBank mRNA | AK028655.1 | - | 2 - 3 | 182 | 88 | |
AK030722.1 | - | 2 - 3 | 183 | 88 | ||
AK043606.1 | - | 2 - 3 | 161 | 88 | ||
AK081915.1 | - | 2 - 3 | 176 | 88 | ||
AK149686.1 | - | 2 - 3 | 171 | 88 | ||
AK166150.1 | - | 2 - 3 | 153 | 88 | ||
BC094437.1 | - | 2 - 3 | 164 | 30682580 | 88 | |
BC117888.1 | - | 2 - 3 | 157 | 40096413 | 88 | |
BC117889.1 | - | 2 - 3 | 160 | 40096414 | 88 | |
BC128069.1 | - | 2 - 3 | 158 | 40096428 | 88 | |
U92793.1 | - | 2 - 3 | 160 | 88 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
