Assay Details
Gene Symbol: Mcm5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK033196.1 | - | 6 - 7 | 811 | 59 | |
AK051795.1 | - | 6 - 7 | 781 | 59 | ||
AK133826.1 | - | 6 - 7 | 812 | 59 | ||
BC090645.1 | - | 6 - 7 | 764 | 59 | ||
BC094416.1 | - | 6 - 7 | 776 | 5707724 | 59 | |
D26090.1 | - | 5 - 6 | 831 | 59 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
