Assay Details
Gene Symbol: Mcm5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK033196.1 | - | 13 - 14 | 1762 | 74 | |
AK051795.1 | - | 13 - 14 | 1732 | 74 | ||
AK088558.1 | - | 6 - 7 | 678 | 74 | ||
AK133826.1 | - | 13 - 14 | 1763 | 74 | ||
AK188730.1 | - | 3 - 4 | 352 | 74 | ||
BC090645.1 | - | 13 - 14 | 1715 | 74 | ||
BC094416.1 | - | 13 - 14 | 1727 | 5707724 | 74 | |
D26090.1 | - | 12 - 13 | 1782 | 74 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
