Assay Details
Gene Symbol: Cebpz | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001024806.2 | NP_001019977.1 | 10 - 11 | 2597 | 97 | |
GenBank mRNA | AK136173.1 | - | 10 - 11 | 2578 | 97 | |
AK145374.1 | - | 10 - 11 | 2579 | 97 | ||
AK155990.1 | - | 10 - 11 | 2580 | 97 | ||
AK158771.1 | - | 10 - 11 | 2574 | 97 | ||
AK159081.1 | - | 10 - 11 | 2594 | 97 | ||
U19891.1 | - | 10 - 11 | 2568 | 97 | ||
U19892.1 | - | 10 - 11 | 2282 | 97 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
