Assay Details
Gene Symbol: Rad51 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011234.4 | NP_035364.1 | 2 - 3 | 308 | 86 | |
GenBank mRNA | AK011242.1 | - | 2 - 3 | 309 | 86 | |
AK045050.1 | - | 2 - 3 | 311 | 86 | ||
AK076468.1 | - | 2 - 3 | 256 | 86 | ||
AK148165.1 | - | 308 | 86 | |||
AK151157.1 | - | 2 - 3 | 287 | 86 | ||
AK151177.1 | - | 2 - 3 | 287 | 86 | ||
AK160282.1 | - | 2 - 3 | 300 | 86 | ||
BC027384.1 | - | 2 - 3 | 273 | 4951015 | 86 | |
D13473.1 | - | 2 - 3 | 263 | 86 | ||
D13803.1 | - | 2 - 3 | 276 | 86 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
