Assay Details
Gene Symbol: Rad51 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011234.4 | NP_035364.1 | 7 - 8 | 864 | 75 | |
GenBank mRNA | AK011242.1 | - | 7 - 8 | 865 | 75 | |
AK076468.1 | - | 7 - 8 | 812 | 75 | ||
AK151157.1 | - | 7 - 8 | 843 | 75 | ||
AK151177.1 | - | 7 - 8 | 843 | 75 | ||
AK160282.1 | - | 7 - 8 | 856 | 75 | ||
BC027384.1 | - | 7 - 8 | 829 | 4951015 | 75 | |
D13473.1 | - | 7 - 8 | 819 | 75 | ||
D13803.1 | - | 7 - 8 | 832 | 75 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
