Assay Details
Gene Symbol: Sin3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001110351.1 | NP_001103821.1 | 19 - 20 | 3451 | 73 | |
NM_011378.2 | NP_035508.2 | 19 - 20 | 3682 | 73 | ||
GenBank mRNA | AK032209.1 | - | 18 - 19 | 3441 | 73 | |
AK136347.1 | - | 13 - 14 | 2376 | 73 | ||
AK136972.1 | - | 4 - 5 | 807 | 73 | ||
BC078454.1 | - | 18 - 19 | 3435 | 73 | ||
L38620.2 | - | 17 - 18 | 3341 | 73 | ||
L38621.2 | - | 17 - 18 | 3341 | 73 | ||
U22394.1 | - | 19 - 20 | 3690 | 73 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
