Assay Details
Gene Symbol: Cdc6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011799.2 | NP_035929.1 | 1 - 2 | 172 | 104 | |
GenBank mRNA | AJ009559.1 | - | 1 - 2 | 189 | 104 | |
AJ223087.1 | - | 1 - 2 | 174 | 104 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
