Assay Details
Gene Symbol: Sh3yl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001289480.1 | NP_001276409.1 | 3 - 4 | 732 | 85 | |
NM_013709.5 | NP_038737.1 | 4 - 5 | 846 | 85 | ||
NR_110343.1 | 5 - 6 | 991 | 85 | |||
GenBank mRNA | AK141199.1 | - | 4 - 5 | 847 | 85 | |
BC010296.1 | - | 3 - 4 | 294 | 3154966 | 85 | |
BC058557.1 | - | 3 - 4 | 709 | 6313231 | 85 | |
BC061488.1 | - | 4 - 5 | 786 | 4483634 | 85 | |
BC110043.1 | - | 4 - 5 | 824 | 5149633 | 85 | |
D85926.1 | - | 4 - 5 | 329 | 85 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
