Assay Details
Gene Symbol: G6pc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001289856.1 | NP_001276785.1 | 4 - 5 | 415 | 96 | |
NM_021331.4 | NP_067306.1 | 4 - 5 | 635 | 96 | ||
GenBank mRNA | AK148465.1 | - | 4 - 5 | 587 | 96 | |
BQ557220.1 | - | 4 - 5 | 415 | 96 | ||
Z47787.1 | - | 4 - 5 | 613 | 96 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2058761 | AK148465.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2058761 | NM_021331.3 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 908490 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 300062 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 950680 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 727874 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 885816 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 176345 | - | - |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P140961 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2058761 | AK148465.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2058761 | NM_021331.3 | mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21307740 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21307745 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21307741 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21307723 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21307726 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21307738 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21307727 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21307754 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21307760 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21307764 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21307734 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21307761 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21307765 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21307752 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21307731 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21307746 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21307732 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21307747 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21307742 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21307758 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21307749 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21307744 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21307736 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21307751 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21307762 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21307721 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21307728 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21307725 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P21307756 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P04555386 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P140961 | - | - |
Whole Mouse Genome G4122A | A_52_P498119 | Z47787.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P498119 | AK148465.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P498119 | BC111905.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P498119 | AK148450.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P498119 | NM_021331.3 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P103227 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P140961 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P01417241 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P01660640 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P01417231 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21307718 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21307758 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21307787 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21307740 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21307704 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21307832 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21307742 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21307779 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21307800 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21307764 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21307697 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21307713 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21307775 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21307831 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21307701 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21307699 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21307749 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21307767 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21307811 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21307783 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21307799 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21307816 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21307752 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21307785 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21307834 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21307761 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21307705 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21307741 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21307807 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21307707 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21307829 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21307708 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21307766 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21307723 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21307791 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21307821 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21307756 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21307817 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21307823 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21307789 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21307825 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21307801 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21307746 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21307725 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21307736 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21307760 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21307726 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21307796 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21307696 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21307717 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21307808 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21307744 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21307747 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21307793 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21307727 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21307782 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21307714 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21307769 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21307778 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21307824 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21307754 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21307813 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21307819 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21307838 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21307788 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21307804 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21307773 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21307729 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21307731 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21307815 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21307745 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21307809 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21307771 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21307751 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21307830 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02151454 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21307736 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21307823 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21307717 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21307825 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02151433 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21307831 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21307741 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02151428 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01826097 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01980645 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21307734 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21307731 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21307840 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21307711 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21307733 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01417228 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01435351 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21307732 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02151437 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21307785 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01560822 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21307775 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21307756 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02134157 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02151435 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01435353 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21307801 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21307749 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21307773 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21307718 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21307747 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21307754 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01364302 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01417231 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21307836 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21307738 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01778906 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02151429 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21307796 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21307725 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21307838 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21307766 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21307746 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21307783 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21307804 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21307708 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21307760 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02151430 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01417233 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21307815 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21307721 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01417230 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21307769 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21307714 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21307779 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21307764 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21307799 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01642689 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21307758 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21307812 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01660639 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21307740 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21307723 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21307771 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21307761 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02151446 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21307821 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21307752 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01758476 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01912340 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21307832 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01980644 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21307778 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10472532 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10472536 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10472534 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10472535 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10472533 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10472537 | - | - |
Affymetrix HT MG-430 PM Array Plate | 1423529_PM_at | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10472531 | - | - |
Mouse Genome 430 2.0 Array | 1423529_at | - | - |
Mouse Genome 430 2.0 Array | 1423529_PM_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 14378_PM_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 14378_at | - | - |
Affymetrix Mouse Clariom S Array | PSR0200007857.mm.1 | TC0200001003.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0200007857.mm.1 | NM_001289856.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0200007857.mm.1 | NM_001289857.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0200007857.mm.1 | NM_021331.4 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0200007859.mm.1 | TC0200001003.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0200007859.mm.1 | NM_001289856.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0200007859.mm.1 | NM_001289857.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0200007859.mm.1 | NM_021331.4 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0200007855.mm.1 | TC0200001003.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0200007855.mm.1 | NM_001289856.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0200007855.mm.1 | NM_001289857.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0200007855.mm.1 | NM_021331.4 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0200007858.mm.1 | TC0200001003.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0200007858.mm.1 | NM_001289856.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0200007858.mm.1 | NM_001289857.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0200007858.mm.1 | NM_021331.4 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0200007856.mm.1 | TC0200001003.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0200007856.mm.1 | NM_001289856.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0200007856.mm.1 | NM_001289857.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0200007856.mm.1 | NM_021331.4 | Refseq |
Mouse Expression Set 430 | 1423529_at | - | - |
Mouse Expression Set 430 | 1423529_PM_at | - | - |
Murine Genome U74v2 Set | 95330_at | - | - |
Murine Genome U74v2 Set | 95330_PM_at | - | - |
Mouse Genome 430A 2.0 Array | 1423529_PM_at | - | - |
Mouse Genome 430A 2.0 Array | 1423529_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1423529_PM_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1423529_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 368388 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
MouseWG-6 v2 Expression BeadChip | 5220070 | - | - |
Illumina mouse-6 v1.0 expression beadchip | 1660114 | - | - |
Illumina mouse-6 v1.0 expression beadchip | 5220070 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0005220070 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0001660114 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2629682 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1234617 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_2629682 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_1234617 | - | - |
mouse-6 v1.1 (Illumina) | scl0003367.1_19-S | - | - |
mouse-6 v1.1 (Illumina) | scl20833.6_119-S | - | - |
Illumina Sentrix mouse-6.v1 bead array | 5220070 | - | - |
Illumina Sentrix mouse-6.v1 bead array | 1660114 | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl0003367.1_19-S | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl20833.6_119-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 1660114 | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 5220070 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_2629682 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_1234617 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1234617 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2629682 | - | - |
