Assay Details
Gene Symbol: Elp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_021448.2 | NP_067423.2 | 9 - 10 | 923 | 108 | |
GenBank mRNA | AF291064.1 | - | 9 - 10 | 889 | 108 | |
AK033246.1 | - | 9 - 10 | 924 | 108 | ||
AK034457.1 | - | 9 - 10 | 922 | 108 | ||
AK050960.1 | - | 9 - 10 | 924 | 108 | ||
AK147826.1 | - | 924 | 108 | |||
AK167725.1 | - | 6 - 7 | 652 | 108 | ||
AK167853.1 | - | 9 - 10 | 922 | 108 | ||
AK168370.1 | - | 9 - 10 | 924 | 108 | ||
AK197756.1 | - | 1 - 2 | 78 | 108 | ||
BC015284.1 | - | 9 - 10 | 915 | 4239056 | 108 | |
BC051943.1 | - | 9 - 10 | 915 | 5686798 | 108 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
