Assay Details
Gene Symbol: Gcsam | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001159297.1 | NP_001152769.1 | 3 - 4 | 238 | 62 | |
NM_008099.4 | NP_032125.3 | 2 - 3 | 172 | 62 | ||
GenBank mRNA | AK036553.1 | - | 2 - 3 | 190 | 62 | |
AY500830.1 | - | 3 - 4 | 146 | 62 | ||
BC099585.1 | - | 2 - 3 | 136 | 30289367 | 62 | |
U13263.1 | - | 2 - 3 | 173 | 62 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
