Assay Details
Gene Symbol: Mycl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK146959.1 | - | 2 - 3 | 582 | 80 | |
BC053059.1 | - | 1 - 2 | 921 | 5704829 | 80 | |
BC089346.1 | - | 2 - 3 | 558 | 6810905 | 80 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
MicroArray Probes:
